Triethoxy(pentafluorophenyl)silane ≥95.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(Triethoxysilyl)pentafluorobenzene, Triethoxy(perfluorophenyl)silane
T3134-1G
T3134-5G
TCIAT3134-1GEA
38
EUR
TCIAT3134-1G
TCIAT3134-5G
Triethoxy(pentafluorophenyl)silane ≥95.0% (by GC)
Triethoxy(pentafluorophenyl)silane
Formula:
C₁₂H₁₅F₅O₃Si Peso molecolare: 330,33 g/mol Punto di ebollizione: 238°C Flash Pt: 104°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD03411257 CAS Number: 20083-34-5 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...