Stampa…

alpha-Tocopherol polyethylene glycol succinate

Fornitore: TCI

T3118-25G T3118-5G
TCIAT3118-25GEA 199 EUR
TCIAT3118-25G TCIAT3118-5G
alpha-Tocopherol polyethylene glycol succinate
alpha-Tocopherol polyethylene glycol succinate
Formula: (C₂H₄O)nC₃₃H₅₄O₅
Punto di fusione: 40°C
CAS Number: 9002-96-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to lump
Solubility in Water almost transparency
Water max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR