Stampa…

3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(9H-Carbazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (9H-Carbazol-3-yl)boronic acid Pinacol ester

T3117-1G T3117-5G
TCIAT3117-1GEA 50 EUR
TCIAT3117-1G TCIAT3117-5G
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole ≥98.0% (by GC)
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Formula: C₁₈H₂₀BNO₂
Peso molecolare: 293,17 g/mol
MDL Number: MFCD16996081
CAS Number: 855738-89-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7336292301538bfcf6856bb70d2eea0b1b7f01b1&vid=7336292301538bfcf6856bb70d2eea0b1b7f01b1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 194.0 to 198.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR