Teniposide ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(5R,5aR,8aR,9S)-9-(((2R,4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-(thiophen-2-yl)hexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one
T3109-20MG
T3109-100MG
TCIAT3109-20MGEA
44
EUR
TCIAT3109-20MG
TCIAT3109-100MG
Teniposide ≥98.0% (by HPLC)
Teniposide
Formula:
C₃₂H₃₂O₁₃S Peso molecolare: 656,66 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00866516 CAS Number: 29767-20-2 EINECS: 249-831-2 Merck Index: 14,09145 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -107.0 to -115.0 deg (C=1, CHCl3:MeOH=9:1) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...