Stampa…
2,4,6-Trichloropyrimidine-5-carbaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 2,4,6-Trichloro-5-formylpyrimidine, 2,4,6-Trichloro-5-pyrimidinecarboxaldehyde
T3100-1G T3100-5G
TCIAT3100-1GEA 20 EUR
TCIAT3100-1G TCIAT3100-5G
2,4,6-Trichloropyrimidine-5-carbaldehyde ≥98.0% (by GC)
2,4,6-Trichloropyrimidine-5-carbaldehyde
Formula: C₅HCl₃N₂O
Peso molecolare: 211,43 g/mol
MDL Number: MFCD02257700
CAS Number: 50270-27-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0a378ea313e7ff54c76fe4033f3de333d246afd&vid=f0a378ea313e7ff54c76fe4033f3de333d246afd&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 130.0 to 134.0 deg-C