Stampa…
4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane ≥97.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)perylene, (3-Perylenyl)boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(perylen-3-yl)-1,3,2-dioxaborolane
T3089-200MG T3089-1G
TCIAT3089-200MGEA 118 EUR
TCIAT3089-200MG TCIAT3089-1G
4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane ≥97.0% (by GC)
4,4,5,5-Tetramethyl-2-(3-perylenyl)-1,3,2-dioxaborolane
Formula: C₂₆H₂₃BO₂
Peso molecolare: 378,28 g/mol
MDL Number: MFCD28098461
CAS Number: 950761-81-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=21cd9c0295bcadbf4904b8ec96ace32e048ab5ba&vid=21cd9c0295bcadbf4904b8ec96ace32e048ab5ba&prid=web&clid=SAPDAM
Visualizza i codici
Purity (GC) min. 97.0 %
Melting point 234.0 to 238.0 deg-C