Stampa…

Tetraethylammonium nitrate ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

T3082-5G T3082-25G
TCIAT3082-5GEA 32 EUR
TCIAT3082-5G TCIAT3082-25G
Tetraethylammonium nitrate ≥98.0% (by titrimetric analysis)
Tetraethylammonium nitrate
Formula: C₈H₂₀N₂O₃
MDL Number: MFCD00041978
CAS Number: 1941-26-0
EINECS: 217-725-5
UN: 1479
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76bcda5270a841e3d48aeadb1df9d8f99b7dfd10&vid=76bcda5270a841e3d48aeadb1df9d8f99b7dfd10&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR