(R,R)-Ts-DENEB®
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Chloro[(R,R)-N-[2-[2-(4-methylbenzyloxy)ethyl]amino-1,2-diphenylethyl]-p-toluenesulphonamide]ruthenium(II), Chloro[(R,R)-N-[2-(4-methylbenzyloxy)ethyl]-N'-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II)
T3078-200MG
T3078-1G
TCIAT3078-200MGEA
43
EUR
TCIAT3078-200MG
TCIAT3078-1G
(R,R)-Ts-DENEB®
(R,R)-Ts-DENEB®
Formula:
C₃₁H₃₃ClN₂O₃RuS Peso molecolare: 650,20 g/mol |
MDL Number:
MFCD20922902 CAS Number: 1333981-84-2 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Brown powder to crystal |
Content (Ruthenium) | 12 to 17 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...