4-(Trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₄H₃F₃N₂ |
MDL Number:
MFCD11226572 CAS Number: 52222-73-8 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...