Tyrphostin 1 ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₁₁H₈N₂O Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00019787 CAS Number: 2826-26-8 UN: 3439 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...