Stampa…

1,3,5-Tris(3-bromophenyl)benzene ≥96.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 3,3''-Dibromo-5'-(3-bromophenyl)-1,1':3',1''-terphenyl

T3073-1G T3073-5G
TCIAT3073-1GEA 65 EUR
TCIAT3073-1G TCIAT3073-5G
1,3,5-Tris(3-bromophenyl)benzene ≥96.0% (by HPLC)
1,3,5-Tris(3-bromophenyl)benzene
Formula: C₂₄H₁₅Br₃
Peso molecolare: 543,10 g/mol
MDL Number: MFCD02323390
CAS Number: 96761-85-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c335c0f1f1fe392fb92461551969bae6a97528f&vid=8c335c0f1f1fe392fb92461551969bae6a97528f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Melting point 170.0 to 174.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR