Stampa…

Tetrachlorophthalonitrile ≥96.0% (by GC, total nitrogen)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3,4,5,6-Tetrachlorobenzene-1,2-dicarbonitrile, Perchlorophthalonitrile

T3067-5G T3067-25G
TCIAT3067-5GEA 26 EUR
TCIAT3067-5G TCIAT3067-25G
Tetrachlorophthalonitrile ≥96.0% (by GC, total nitrogen)
Tetrachlorophthalonitrile
Formula: C₈Cl₄N₂
Peso molecolare: 265,91 g/mol
MDL Number: MFCD00019740
CAS Number: 1953-99-7
EINECS: 217-783-1
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=feb4a1a017f50071efdb5ead47419fabea4c6a8e&vid=feb4a1a017f50071efdb5ead47419fabea4c6a8e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (with Total Nitrogen) min. 96.0 %
Melting point 249.0 to 253.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR