Stampa…

2-(2-Thienyl)ethyl-p-toluenesulphonate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-(Thiophen-2-yl)ethyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid 2-(2-Thienyl)ethyl Ester

T3064-5G T3064-25G
TCIAT3064-5GEA 64 EUR
TCIAT3064-5G TCIAT3064-25G
2-(2-Thienyl)ethyl-p-toluenesulphonate ≥95.0% (by GC)
2-(2-Thienyl)ethyl-p-toluenesulphonate
Formula: C₁₃H₁₄O₃S₂
Peso molecolare: 282,37 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD05150366
CAS Number: 40412-06-4
EINECS: 254-911-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3457483d33ec38139d83f0dd323bf0a0e8c04f3f&vid=3457483d33ec38139d83f0dd323bf0a0e8c04f3f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 30.0 to 34.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR