Stampa…

Tinidazole ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1-[2-(Ethylsulphonyl)ethyl]-2-methyl-5-nitroimidazole, 1-(2-(Ethylsulphonyl)ethyl)-2-methyl-5-nitro-1H-imidazole

T3058-5G T3058-25G
TCIAT3058-5GEA 16 EUR
TCIAT3058-5G TCIAT3058-25G
Tinidazole ≥98.0% (by HPLC, titration analysis)
Tinidazole
Formula: C₈H₁₃N₃O₄S
Peso molecolare: 247,27 g/mol
MDL Number: MFCD00057217
CAS Number: 19387-91-8
EINECS: 243-014-4
Merck Index: 14,09447
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11097366018dc26b8ad9657983d048e762048289&vid=11097366018dc26b8ad9657983d048e762048289&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR