1,1,2,2-Tetrafluoroethyl-2,2,2-trifluoroethyl ether ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1,1,2,2-Tetrafluoroethyl 2,2,2-trifluoroethyl ether
T3057-5G
T3057-25G
TCIAT3057-5GEA
31
EUR
TCIAT3057-5G
TCIAT3057-25G
1,1,2,2-Tetrafluoroethyl-2,2,2-trifluoroethyl ether ≥99.0% (by GC)
1,1,2,2-Tetrafluoroethyl-2,2,2-trifluoroethyl ether
Formula:
C₄H₃F₇O Peso molecolare: 200,06 g/mol Punto di ebollizione: 56°C |
MDL Number:
MFCD00236118 CAS Number: 406-78-0 UN: 3271 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...