Stampa…

Tris[4-(2-thienyl)phenyl]amine ≥98.0% (by GC)

Fornitore: TCI EUROPE

T3050-1G T3050-5G
TCIAT3050-1GEA 137 EUR
TCIAT3050-1G TCIAT3050-5G
Tris[4-(2-thienyl)phenyl]amine ≥98.0% (by GC)
Tris[4-(2-thienyl)phenyl]amine
Formula: C₃₀H₂₁NS₃
Peso molecolare: 491,69 g/mol
CAS Number: 142807-63-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd50b56644b88f783b87396e38fd0aaedfdd0616&vid=fd50b56644b88f783b87396e38fd0aaedfdd0616&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR