3-(Trimethylsilyl)propiolic acid ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(Trimethylsilyl)propynoic acid
T3040-1G
T3040-5G
TCIAT3040-1GEA
274
EUR
TCIAT3040-1G
TCIAT3040-5G
3-(Trimethylsilyl)propiolic acid ≥97.0% (by GC, titration analysis)
3-(Trimethylsilyl)propiolic acid
Formula:
C₆H₁₀O₂Si Peso molecolare: 142,23 g/mol Punto di ebollizione: 110°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00190216 CAS Number: 5683-31-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 46.0 to 50.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...