1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
TIPS-EBX, 1-((Tris(1-methylethyl)silyl)ethynyl)-1,2-benziodoxol-3(1H)-one
T3039-200MG
T3039-1G
TCIAT3039-200MGEA
47
EUR
TCIAT3039-200MG
TCIAT3039-1G
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one ≥98.0% (by HPLC)
1-[(Triisopropylsilyl)ethynyl]-1,2-benziodoxol-3(1H)-one
Formula:
C₁₈H₂₅IO₂Si Peso molecolare: 428,39 g/mol Punto di fusione: 170°C |
MDL Number:
MFCD18632570 CAS Number: 181934-30-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...