Stampa…

Trimethyl-1,3,5-cyclohexanetricarboxylate (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,3,5-Cyclohexanetricarboxylic Acid Trimethyl Ester

T3033-1G T3033-5G
TCIAT3033-1GEA 163 EUR
TCIAT3033-1G TCIAT3033-5G
Trimethyl-1,3,5-cyclohexanetricarboxylate (cis and trans mixture) cis- and trans-mixture ≥98.0% (by GC)
Trimethyl-1,3,5-cyclohexanetricarboxylate (cis and trans mixture)
Formula: C₁₂H₁₈O₆
Peso molecolare: 258,27 g/mol
Punto di fusione: 46°C
CAS Number: 52831-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f0693e6c5ea9b1cdcc7cbbdafea7dfb189a8479&vid=5f0693e6c5ea9b1cdcc7cbbdafea7dfb189a8479&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR