Tiotropium bromide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Synonyms:
rel-(1R,2R,4S,5S,7s)-7-(2-Hydroxy-2,2-di(thiophen-2-yl)acetoxy)-9,9-dimethyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-9-ium bromide
Formula:
C₁₉H₂₂BrNO₄S₂ Peso molecolare: 472.41 g/mol Punto di fusione: 230°C |
MDL Number:
MFCD00867027 CAS Number: 136310-93-5 Merck Index: 14,09454 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...