Trichloro[3-(pentafluorophenyl)propyl]silane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
T3030-1G
T3030-5G
TCIAT3030-1GEA
57
EUR
TCIAT3030-1G
TCIAT3030-5G
Trichloro[3-(pentafluorophenyl)propyl]silane ≥98.0% (by GC)
Trichloro[3-(pentafluorophenyl)propyl]silane
Formula:
C₉H₆Cl₃F₅Si |
MDL Number:
MFCD01074474 CAS Number: 78900-02-4 UN: 2987 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...