Stampa…

Tris(3,5-dimethyl-1-pyrazolyl)methane ≥98.0% (by GC)

Fornitore: TCI EUROPE

T3025-200MG T3025-1G
TCIAT3025-200MGEA 33 EUR
TCIAT3025-200MG TCIAT3025-1G
Tris(3,5-dimethyl-1-pyrazolyl)methane ≥98.0% (by GC)
Tris(3,5-dimethyl-1-pyrazolyl)methane
Formula: C₁₆H₂₂N₆
Peso molecolare: 298,39 g/mol
CAS Number: 28791-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a1b1e2f14b357ad42e0e59e22b40c232917b6eb&vid=1a1b1e2f14b357ad42e0e59e22b40c232917b6eb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR