4-(1H-1,2,4-Triazol-1-yl)aniline ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-(1,2,4-Triazol-1-yl)aniline, 1-(4-Aminophenyl)-1,2,4-triazole
T3013-200MG
T3013-1G
TCIAT3013-200MGEA
52
EUR
TCIAT3013-200MG
TCIAT3013-1G
4-(1H-1,2,4-Triazol-1-yl)aniline ≥98.0% (by GC, titration analysis)
4-(1H-1,2,4-Triazol-1-yl)aniline
Formula:
C₈H₈N₄ Peso molecolare: 160,1759 g/mol |
MDL Number:
MFCD00171470 CAS Number: 6523-49-5 |
Risultati del test di specificazione
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 140.0 to 144.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...