4-(m-Tolylamino)pyridine-3-sulphonamide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
4-(m-Tolylamino)-3-sulphamoylpyridine, 4-(3-Methylphenyl)aminopyridine-3-sulphonamide, Torsemide Related Compound A, 4-(3-Methylphenyl)amino-3-sulphamoylpyridine
T3007-5G
T3007-25G
TCIAT3007-5GEA
72
EUR
TCIAT3007-5G
TCIAT3007-25G
4-(m-Tolylamino)pyridine-3-sulphonamide ≥98.0% (by HPLC, titration analysis)
4-(m-Tolylamino)pyridine-3-sulphonamide
Formula:
C₁₂H₁₃N₃O₂S Peso molecolare: 263,32 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00661332 CAS Number: 72811-73-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 169.0 to 173.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...