Stampa…

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: TBTA, Tris((1-benzyl-1H-1,2,3-triazol-4-yl)methyl)amine

T2993-1G T2993-5G
TCIAT2993-1GEA 115 EUR
TCIAT2993-1G TCIAT2993-5G
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine ≥97.0% (by HPLC, titration analysis)
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Formula: C₃₀H₃₀N₁₀
Peso molecolare: 530,64 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD09265124
CAS Number: 510758-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01e6c7b918e1e48e634021bc5a9d26238abc7072&vid=01e6c7b918e1e48e634021bc5a9d26238abc7072&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystaline
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR