Tebuconazole ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-(4-Chlorophenyl)-4,4-dimethyl-3-(1H-1,2,4-triazol-1-ylmethyl)-3-pentanol
T2988-1G
T2988-5G
TCIAT2988-1GEA
36
EUR
TCIAT2988-1G
TCIAT2988-5G
Tebuconazole ≥98.0% (by HPLC, titration analysis)
Tebuconazole
Formula:
C₁₆H₂₂ClN₃O Peso molecolare: 307,82 g/mol |
MDL Number:
MFCD02674797 CAS Number: 107534-96-3 EINECS: 403-640-2 UN: 3077 ADR: 9,III Merck Index: 14,09092 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 101.0 to 105.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...