Trifluoromethanesulphonanilide ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
T2985-1G
T2985-5G
TCIAT2985-1GEA
86
EUR
TCIAT2985-1G
TCIAT2985-5G
Trifluoromethanesulphonanilide ≥98.0% (by HPLC, titration analysis)
Trifluoromethanesulphonanilide
Formula:
C₇H₆F₃NO₂S |
MDL Number:
MFCD00521820 CAS Number: 456-64-4 UN: 2811 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 65.0 to 69.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...