Stampa…

4-(Trifluoromethyl)thiophenol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-Mercaptobenzotrifluoride, 4-(Trifluoromethyl)benzene-1-thiol, 4-(Trifluoromethyl)benzenethiol

T2972-1G T2972-5G
TCIAT2972-1GEA 46 EUR
TCIAT2972-1G TCIAT2972-5G
4-(Trifluoromethyl)thiophenol ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)thiophenol
Formula: C₇H₅F₃S
Peso molecolare: 178,18 g/mol
MDL Number: MFCD00042320
CAS Number: 825-83-2
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d328dac36298f713bf4488fd89e708d500d7a0c5&vid=d328dac36298f713bf4488fd89e708d500d7a0c5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR