Tetrakis(4-bromophenyl)methane ≥95.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₂₅H₁₆Br₄ Punto di fusione: 300°C |
MDL Number:
MFCD19688472 CAS Number: 105309-59-9 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...