Tripyrrolidinophosphine oxide ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
Tri(pyrrolidin-1-yl)phosphine oxide
Formula:
C₁₂H₂₄N₃OP Peso molecolare: 257.32 g/mol Punto di ebollizione: 133°C |
MDL Number:
MFCD00014095 CAS Number: 6415-07-2 EINECS: 229-118-2 |
|
Risultati del test di specificazione
Appearance | Colorless to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...