Stampa…

2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(Bromomethyl)phenylboronic acid Pinacol ester, 4-Bromomethylphenylboronic acid, pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide, 2-[4-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T2945-1G T2945-5G
TCIAT2945-1GEA 22 EUR
TCIAT2945-1G TCIAT2945-5G
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₈BBrO₂
Peso molecolare: 297,00 g/mol
MDL Number: MFCD02179493
CAS Number: 138500-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6073fe6ec4f69b23e24c3242996b4a7394d95f2a&vid=6073fe6ec4f69b23e24c3242996b4a7394d95f2a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 84.0 to 88.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR