Stampa…

1,2,3,9-Tetrahydro-9-methyl-4H-carbazole-4-one ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1,2,3,4-Tetrahydro-9-methyl-4-oxocarbazole, 1,2,3,4-Tetrahydro-9-methylcarbazol-4-one, Ondansetron Related Compound C

T2942-5G T2942-25G
TCIAT2942-5GEA 17 EUR
TCIAT2942-5G TCIAT2942-25G
1,2,3,9-Tetrahydro-9-methyl-4H-carbazole-4-one ≥98.0% (by GC)
1,2,3,9-Tetrahydro-9-methyl-4H-carbazole-4-one
Formula: C₁₃H₁₃NO
Peso molecolare: 199,2500 g/mol
MDL Number: MFCD00173748
CAS Number: 27387-31-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26205b4865c962514cd6a991215207f3d4d855e8&vid=26205b4865c962514cd6a991215207f3d4d855e8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 199.0 to 203.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR