Tin(IV) 2,3-naphthalocyanine dichloride
Fornitore: TCI
|
Pericolo
|
Formula:
C₄₈H₂₄Cl₂N₈Sn |
MDL Number:
MFCD00171716 CAS Number: 26857-61-4 UN: 3146 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | Green to Dark green powder to crystal |
Elemental analysis (Carbon) | 60.70 to 67.10 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...