3-(3-Trifluoromethylphenyl)propionic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(3-(Trifluoromethyl)phenyl)propanoic acid, 3-[3-(Trifluoromethyl)phenyl]propionic acid, 3-(Trifluoromethyl)hydrocinnamic acid
T2931-1G
T2931-5G
TCIAT2931-1GEA
17
EUR
TCIAT2931-1G
TCIAT2931-5G
3-(3-Trifluoromethylphenyl)propionic acid ≥98.0% (by GC, titration analysis)
3-(3-Trifluoromethylphenyl)propionic acid
Formula:
C₁₀H₉F₃O₂ Peso molecolare: 218,18 g/mol Punto di ebollizione: 149°C Punto di fusione: 35°C |
MDL Number:
MFCD00090487 CAS Number: 585-50-2 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...