Stampa…

1-(p-Toluenesulphonyl)-3-nitro-1,2,4-triazole ≥98.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-Nitro-1-tosyl-1,2,4-triazole, 3-Nitro-1-(p-toluenesulphonyl)-1,2,4-triazole, 3-Nitro-1-tosyl-1H-1,2,4-triazole

T2930-1G T2930-5G
TCIAT2930-1GEA 18 EUR
TCIAT2930-1G TCIAT2930-5G
1-(p-Toluenesulphonyl)-3-nitro-1,2,4-triazole ≥98.0% (by HPLC)
1-(p-Toluenesulphonyl)-3-nitro-1,2,4-triazole
Formula: C₉H₈N₄O₄S
Peso molecolare: 268,25 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00009756
CAS Number: 77451-51-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=179e7c988962f47e910012a7810b737c4c55599f&vid=179e7c988962f47e910012a7810b737c4c55599f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 20.40 to 21.10 %
Melting point 132.0 to 136.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR