Stampa…

4-(Trifluoromethyl)hydrocinnamic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 3-[4-(Trifluoromethyl)phenyl]propanoic acid

T2927-1G T2927-5G
TCIAT2927-1GEA 18 EUR
TCIAT2927-1G TCIAT2927-5G
4-(Trifluoromethyl)hydrocinnamic acid ≥98.0% (by titrimetric analysis)
4-(Trifluoromethyl)hydrocinnamic acid
Formula: C₁₀H₉F₃O₂
MDL Number: MFCD00674032
CAS Number: 53473-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c694a6174300806b5662416db26ec0818d3d6567&vid=c694a6174300806b5662416db26ec0818d3d6567&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR