(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(R)-1,2,3,4-Tetrahydronaphthalen-1-amine
T2926-5G
T2926-25G
TCIAT2926-5GEA
63
EUR
TCIAT2926-5G
TCIAT2926-25G
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine ≥98.0% (by GC, titration analysis)
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine
Formula:
C₁₀H₁₃N Peso molecolare: 147,22 g/mol Punto di ebollizione: 70°C |
MDL Number:
MFCD00671629 CAS Number: 23357-46-2 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (LC) | min. 98.0 ee% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...