Stampa…

4-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC)

Fornitore: TCI

Synonyms: 4-Formylphenylboronic acid pinacol ester, 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde, 4-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

T2923-1G T2923-5G
TCIAT2923-1GEA 22 EUR
TCIAT2923-1G TCIAT2923-5G
4-Formylbenzeneboronic acid pinacol ester ≥97.0% (by GC)
4-Formylbenzeneboronic acid pinacol ester
Formula: C₁₃H₁₇BO₃
MDL Number: MFCD04972375
CAS Number: 128376-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bbf5b5749a6ae596f855ff1379c7294220ca550&vid=9bbf5b5749a6ae596f855ff1379c7294220ca550&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 59.0 to 63.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR