Stampa…

1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1,2,3,4-Tetrahydro-1-naphthoic acid

T2903-5G T2903-25G
TCIAT2903-5GEA 72 EUR
TCIAT2903-5G TCIAT2903-25G
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid ≥98.0% (by GC, titration analysis)
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid
Formula: C₁₁H₁₂O₂
Punto di ebollizione: 135°C
MDL Number: MFCD02179127
CAS Number: 1914-65-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a984828b3e891f94c2e288c536407b4a82412bb9&vid=a984828b3e891f94c2e288c536407b4a82412bb9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 82.0 to 86.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR