Stampa…

Tris[2-(dimethylamino)ethyl]amine ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Me6TREN

T2898-1G T2898-5G
TCIAT2898-1GEA 55 EUR
TCIAT2898-1G TCIAT2898-5G
Tris[2-(dimethylamino)ethyl]amine ≥98.0% (by GC)
Tris[2-(dimethylamino)ethyl]amine
Formula: C₁₂H₃₀N₄
Peso molecolare: 230,40 g/mol
MDL Number: MFCD00015607
CAS Number: 33527-91-2

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR