4-Methylphenylboronic acid pinacol ester ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4,5,5-Tetramethyl-2-(p-tolyl)-1,3,2-dioxaborolane
T2888-1G
T2888-5G
TCIAT2888-1GEA
24
EUR
TCIAT2888-1G
TCIAT2888-5G
4-Methylphenylboronic acid pinacol ester ≥98.0% (by GC)
4-Methylphenylboronic acid pinacol ester
Formula:
C₁₃H₁₉BO₂ Peso molecolare: 218,1 g/mol |
MDL Number:
MFCD03453662 CAS Number: 195062-57-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 52.0 to 56.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...