(Trifluoromethyl)tris(triphenylphosphine)copper(I) ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
T2883-1G
T2883-5G
TCIAT2883-1GEA
241
EUR
TCIAT2883-1G
TCIAT2883-5G
(Trifluoromethyl)tris(triphenylphosphine)copper(I) ≥98.0% (by titrimetric analysis)
(Trifluoromethyl)tris(triphenylphosphine)copper(I)
Formula:
C₅₅H₄₈CuF₃P₃ |
MDL Number:
MFCD22200540 CAS Number: 325810-07-9 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Chelometric Titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 7.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...