1,3,5-Tri(2-thienyl)benzene ≥98.0% (by GC)
Fornitore: TCI EUROPE
T2866-5G
T2866-1G
TCIAT2866-5GEA
293
EUR
TCIAT2866-5G
TCIAT2866-1G
1,3,5-Tri(2-thienyl)benzene ≥98.0% (by GC)
1,3,5-Tri(2-thienyl)benzene
Formula:
C₁₈H₁₂S₃ Peso molecolare: 324,47 g/mol Punto di ebollizione: 298°C |
MDL Number:
MFCD02323391 CAS Number: 15509-95-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 157.0 to 161.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...