Trichloro(6-phenylhexyl)silane ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(6-Phenylhexyl)trichlorosilane
T2854-5G
T2854-25G
TCIAT2854-5GEA
110
EUR
TCIAT2854-5G
TCIAT2854-25G
Trichloro(6-phenylhexyl)silane ≥98.0% (by GC)
Trichloro(6-phenylhexyl)silane
Formula:
C₁₂H₁₇Cl₃Si Peso molecolare: 295,70 g/mol Punto di ebollizione: 92°C |
MDL Number:
MFCD09953885 CAS Number: 18035-33-1 UN: 2987 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...