Stampa…

Toremifene citrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (Z)-4-Chloro-1-(4-dimethylaminoethoxyphenyl)-1,2-diphenyl-1-butene Citrate, (Z)-2-[4-(4-Chloro-1,2-diphenyl-1-butenyl)phenoxy]-N,N-dimethylethanamine Citrate

T2832-5G T2832-1G
TCIAT2832-5GEA 325 EUR
TCIAT2832-5G TCIAT2832-1G
Toremifene citrate ≥98.0% (by HPLC, titration analysis)
Toremifene citrate
Formula: C₂₆H₂₈ClNO
Peso molecolare: 598,09 g/mol
Punto di fusione: 162°C
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD01729016
CAS Number: 89778-27-8
Merck Index: 14,09550
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11eb21f9c9a39a2b63b8f046903ca5a4908bcac1&vid=11eb21f9c9a39a2b63b8f046903ca5a4908bcac1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR