Stampa…

Tetracaine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 4-(Butylamino)benzoic Acid 2-(Dimethylamino)ethyl Ester, 2-(Dimethylamino)ethyl 4-(Butylamino)benzoate

T2789-5G T2789-25G
TCIAT2789-5GEA 39 EUR
TCIAT2789-5G TCIAT2789-25G
Tetracaine ≥98.0% (by GC, titration analysis)
Tetracaine
Formula: C₁₅H₂₄N₂O₂
Peso molecolare: 264,37 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00053787
CAS Number: 94-24-6
EINECS: 202-316-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=edea5de8ca3c1badd4a7c64243c5a831e42672ad&vid=edea5de8ca3c1badd4a7c64243c5a831e42672ad&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 41.0 to 45.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR