Truxene ≥98.0% (by HPLC)
Fornitore: TCI
Synonyms:
10,15-Dihydro-5H-tribenzo[a,f,k]trindene, Tribenzylenebenzene, 10,15-Dihydro-5H-diindeno[1,2-a:1',2'-c]fluorene
Formula:
C₂₇H₁₈ Peso molecolare: 342.44 g/mol Punto di fusione: 370°C |
MDL Number:
MFCD00021180 CAS Number: 548-35-6 EINECS: 208-944-7 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...