Tamsulosin hydrochloride ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-[(R)-2-[2-(2-Ethoxyphenoxy)ethylamino]propyl]]-2-methoxybenzenesulphonamide Hydrochloride
T2749-1G
T2749-100MG
TCIAT2749-1GEA
328
EUR
TCIAT2749-1G
TCIAT2749-100MG
Tamsulosin hydrochloride ≥98.0% (by HPLC, titration analysis)
Tamsulosin hydrochloride
Formula:
C₂₀H₂₈N₂O₅S Peso molecolare: 444,97 g/mol Punto di fusione: 230°C |
MDL Number:
MFCD00922997 CAS Number: 106463-17-6 Merck Index: 14,09049 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -17.5 to -20.5 deg (C=1, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...