Teprenone [mixture of (5E,9E,13E)- and (5Z,9E,13E)- isomers] ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
6,10,14,18-Tetramethyl-5,9,13,17-nonadecatetraen-2-one, Geranylgeranylacetone
T2701-1G
T2701-5G
TCIAT2701-1GEA
85
EUR
TCIAT2701-1G
TCIAT2701-5G
Teprenone [mixture of (5E,9E,13E)- and (5Z,9E,13E)- isomers] ≥95.0% (by GC)
Teprenone [mixture of (5E,9E,13E)- and (5Z,9E,13E)- isomers]
Formula:
C₂₃H₃₈O Peso molecolare: 330,56 g/mol Punto di ebollizione: 160°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00869547 CAS Number: 6809-52-5 Merck Index: 14,09228 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % (total of isomer) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...