Stampa…

Tetrakis(2,4-pentanedionato)titanium(IV) ∼63% in isopropanol (IPA)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Titanium(IV) Acetylacetonate, Acetylacetone Titanium(IV) Salt

T2610-25G T2610-500G
TCIAT2610-25GEA 16 EUR
TCIAT2610-25G TCIAT2610-500G
Tetrakis(2,4-pentanedionato)titanium(IV) ∼63% in isopropanol (IPA)
Tetrakis(2,4-pentanedionato)titanium(IV)
Formula: C₂₀H₂₈O₈Ti
Peso molecolare: 444,30 g/mol
MDL Number: MFCD00045004
CAS Number: 17501-79-0
EINECS: 241-511-0
UN: 1219
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange clear liquid
Concentration (Gravimetric Method) 63.0 to 68.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR